missing translation for 'onlineSavingsMsg'
Learn More

1,8-Dihydroxyanthraquinone, 95%

Product Code. p-111953 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
100 g
500 g
Packaging:
Glass bottle
Unit Size:
100g
500g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10774601 5 g Glass bottle 5g
10244570 100 g Glass bottle 100g
10477870 500 g Glass bottle 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10774601 Supplier Thermo Scientific Acros Supplier No. 114830050
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 117-10-2 | C14H8O4 | 240.21 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 117-10-2
Molecular Formula C14H8O4
Molecular Weight (g/mol) 240.21
MDL Number MFCD00001211
InChI Key QBPFLULOKWLNNW-UHFFFAOYSA-N
Synonym danthron, 1,8-dihydroxyanthraquinone, chrysazin, dantron, antrapurol, laxanthreen, diaquone, istizin, laxanorm, altan
PubChem CID 2950
ChEBI CHEBI:3682
IUPAC Name 1,8-dihydroxyanthracene-9,10-dione
SMILES C1=CC2=C(C(=C1)O)C(=O)C3=C(C2=O)C=CC=C3O

Specifications

CAS Min % 90.0
CAS Max % 100.0
Melting Point 190.0°C to 195.0°C
Color Brown or Brown-Orange
Flash Point >200°C
Infrared Spectrum Authentic
Assay Percent Range 94% min. (HPLC)
Packaging Glass bottle
Quantity 5 g
Beilstein 08, 458
Merck Index 15, 2814
Solubility Information Solubility in water: insoluble. Other solubilities: almost insoluble in alcohol,moderately soluble in ether,chloroform,soluble in 10 parts hot glacial acetic acid
Formula Weight 240.21
Percent Purity 95%
Physical Form Powder
Chemical Name or Material 1, 8-Dihydroxyanthraquinone, 90-95%
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 1,8-Dihydroxyanthraquinone
Signal Word
  • Warning
Hazard Category
  • Carcinogenicity Category 2
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H319-Causes serious eye irritation.
  • H351-Suspected of causing cancer.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contain: 1,8-Dihydroxyanthraquinone

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.