missing translation for 'onlineSavingsMsg'
Learn More

1,8-Diazabicyclo[5.4.0]undec-7-ene, Sigma-Aldrich™

Product Code. 15878990
Change view
Click to view available options
Quantity:
25 mL
100 mL
Unit Size:
100mL
25mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15878990 100 mL 100mL
15868990 25 mL 25mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15878990 Supplier Sigma Aldrich Supplier No. 8032820100
 Restricted Item
Estimated Shipment: 16-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

For synthesis

We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product has been enhanced for catalytic efficiency.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

Grade Research
Molecular Weight (g/mol) 152.24

Specifications

Melting Point <-70°C
Density 1.02 g/cm3 at 20°C
pH 12.8 (20°C, 10 g/L in H2O)
MDL Number MFCD00006930
Quantity 100 mL
Synonym 1,8-Diazabicyclo[5.4.0]undec-7-ene, 1,8-Diazabicyclo[5.4.0]undec-7-ene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.