Learn More
1,8-Diaminonaphthalene, TRC
Product Code. 30351645
Change view
Click to view available options
Quantity:
25 g
100 g
250 g
Unit Size:
100g
250g
25g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30351645 250 g 250g
30353383 25 g 25g
30357353 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30351645 Supplier Toronto Research Chemicals Supplier No. TRCD416445250G
 Restricted Item
Estimated Shipment: 19-08-2026
Add to basket
Add to basket
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 479-27-6
Molecular Formula C10 H10 N2
Molecular Weight (g/mol) 158.2
Synonym 1,8-Diamine naphthaline, 1,8-Diaminonaphthalene, 1,8-Naphthylenediamine, NSC 6081
IUPAC Name naphthalene-1,8-diamine
SMILES Nc1cccc2cccc(N)c12
Specifications
Quantity 250 g
InChI Formula InChI=1S/C10H10N2/c11-8-5-1-3-7-4-2-6-9(12)10(7)8/h1-6H,11-12H2
Formula Weight 158.0844
Chemical Name or Material 1,8-Diaminonaphthalene