Learn More
1,7-Dichloro-octamethyltetrasiloxane, TRC
Specifications
Specifications
| Chemical Name or Material | 1,7-Dichloro-octamethyltetrasiloxane |
| CAS | 2474-02-4 |
| Molecular Formula | C8H24Cl2O3Si4 |
| Quantity | 100 mg |
| InChI Formula | InChI=1S/C8H24Cl2O3Si4/c1-14(2,9)11-16(5,6)13-17(7,8)12-15(3,4)10/h1-8H3 |
| SMILES | C[Si](C)(Cl)O[Si](C)(C)O[Si](C)(C)O[Si](C)(C)Cl |
| IUPAC Name | chloro-[[[chloro(dimethyl)silyl]oxy-dimethylsilyl]oxy-dimethylsilyl]oxy-dimethylsilane |
| Molecular Weight (g/mol) | 351.52 |
| Formula Weight | 350.018 |