Learn More
1,7,7-Trimethylbicyclo[2.2.1]heptane-2,3-dione, 98%, BLD Pharm™
Specifications
Specifications
| CAS | 10373-78-1 |
| Molecular Formula | C10H14O2 |
| MDL Number | MFCD00064160 |
| Quantity | 500 g |
| SMILES | O=C1C(C2(C)C)(C)CCC2C1=O |
| Molecular Weight (g/mol) | 166.22 |
| Percent Purity | 98% |
| Chemical Name or Material | 1,7,7-Trimethylbicyclo[2.2.1]heptane-2,3-dione |