Learn More
1-[[(6R,7R)-7-Amino-2-carboxy-8-oxo-5-thia-1-azabicyclo[4.2.0]oct-2-en-3-yl]methyl]-1-methylpyrrolidinium Chloride, TRC
Product Code. 30311820
Change view
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30311820 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30311820 Supplier Toronto Research Chemicals Supplier No. TRCA603160250MG
 Restricted Item
Estimated Shipment: 08-10-2026
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material 1-{[(6R,7R)-7-Amino-2-carboxy-8-oxo-5-thia-1-azabicyclo[4.2.0]oct-2-en-3-yl]methyl}-1-methylpyrrolidin-1-ium chloride
CAS 103121-85-3
Molecular Formula C13 H20 N3 O3 S . Cl
Quantity 250 mg
Synonym Pyrrolidinium, 1-[[(6R,7R)-7-amino-2-carboxy-8-oxo-5-thia-1-azabicyclo[4.2.0]oct-2-en-3-yl]methyl]-1-methyl-, chloride, Pyrrolidinium, 1-[(7-amino-2-carboxy-8-oxo-5-thia-1-azabicyclo[4.2.0]oct-2-en-3-yl)methyl]-1-methyl-, hydroxide, inner salt, monohydrochloride, (6R-trans)-, 1-[[(6R,7R)-7-Amino-2-carboxy-8-oxo-5-thia-1-azabicyclo[4.2.0]oct-2-en-3-yl]methyl]-1-methylpyrrolidinium chloride
InChI Formula InChI=1S/C13H19N3O3S.ClH/c1-16(4-2-3-5-16)6-8-7-20-12-9(14)11(17)15(12)10(8)13(18)19;/h9,12H,2-7,14H2,1H3;1H/t9-,12-;/m1./s1
SMILES [Cl-].C[N+]1(CC2=C(N3[C@H](SC2)[C@H](N)C3=O)C(=O)O)CCCC1
IUPAC Name (6R,7R)-7-amino-3-[(1-methylpyrrolidin-1-ium-1-yl)methyl]-8-oxo-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylic acid;chloride
Molecular Weight (g/mol) 333.83
Formula Weight 333.0914