Learn More
1,6-Dimethoxyhexane, 97%, Thermo Scientific™
Product Code. p-7028019
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
Product Code. 11476177
Brand: Thermo Scientific Alfa Aesar B25130.06
Request Bulk or Custom Format
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 13179-98-1
Molecular Formula C8H18O2
Molecular Weight (g/mol) 146.23
MDL Number MFCD00142569
InChI Key GZHQUHIMIVEQFV-UHFFFAOYSA-N
PubChem CID 542359
IUPAC Name 1,6-dimethoxyhexane
SMILES COCCCCCCOC
Specifications
Density 0.851
Boiling Point 182°C to 183°C
Flash Point 69°C (157°F)
Refractive Index 1.415
Quantity 5 g
Synonym hexane,1,6-dimethoxy, hexamethylene glycol dimethyl ether, hexane, 1,6-dimethoxy, acmc-1c0t4, 1,6-dimethoxyhexane
Formula Weight 146.23
Percent Purity 97%
Chemical Name or Material 1,6-Dimethoxyhexane