CAS RN 13179-98-1
CAS RN 13179-98-1
IUPAC Name:
1,6-dimethoxyhexane
Synonyms:
1,6-dimethoxyhexane
Molecular Weight (g/mol):
146.23
Molecular Formula:
C8H18O2
InChi Key:
GZHQUHIMIVEQFV-UHFFFAOYSA-N
SMILES:
COCCCCCCOC
1
–
1
of
1
results
1
1,6-Dimethoxyhexane, 97%, Thermo Scientific™
CAS: 13179-98-1 Molecular Formula: C8H18O2 Molecular Weight (g/mol): 146.23 MDL Number: MFCD00142569 InChI Key: GZHQUHIMIVEQFV-UHFFFAOYSA-N PubChem CID: 542359 IUPAC Name: 1,6-dimethoxyhexane SMILES: COCCCCCCOC