Learn More
1-[(6-Aminohexyl)sulfanyl]ethan-1-one Hydrochloride, TRC
Specifications
Specifications
| Chemical Name or Material | 1-[(6-Aminohexyl)sulfanyl]ethan-1-one Hydrochloride |
| CAS | 98486-87-4 |
| Molecular Formula | C8H18ClNOS |
| Quantity | 10 mg |
| Synonym | 1-[(6-Aminohexyl)sulfanyl]ethan-1-one Hydrochloride |
| InChI Formula | InChI=1S/C8H17NOS.ClH/c1-8(10)11-7-5-3-2-4-6-9;/h2-7,9H2,1H3;1H |
| SMILES | CC(SCCCCCCN)=O.Cl |
| IUPAC Name | S-(6-aminohexyl) ethanethioate hydrochloride |
| Molecular Weight (g/mol) | 211.75 |
| Formula Weight | 211.09 |