Learn More
1-[(6-Aminohexyl)sulfanyl]ethan-1-one Hydrochloride, TRC
Product Code. 30344526
Change view
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30344526 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30344526 Supplier Toronto Research Chemicals Supplier No. TRCA63924350MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Chemical Name or Material 1-[(6-Aminohexyl)sulfanyl]ethan-1-one Hydrochloride
CAS 98486-87-4
Molecular Formula C8H18ClNOS
Quantity 50 mg
Synonym 1-[(6-Aminohexyl)sulfanyl]ethan-1-one Hydrochloride
InChI Formula InChI=1S/C8H17NOS.ClH/c1-8(10)11-7-5-3-2-4-6-9;/h2-7,9H2,1H3;1H
SMILES CC(SCCCCCCN)=O.Cl
IUPAC Name S-(6-aminohexyl) ethanethioate hydrochloride
Molecular Weight (g/mol) 211.75
Formula Weight 211.09
Produkttitel
Vælg et problem
Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.