Learn More
1,6,7,12-Tetrachloroperylene Tetracarboxylic Acid Dianhydride, TRC
Specifications
Specifications
| Chemical Name or Material | 1,6,7,12-Tetrachloroperylene Tetracarboxylic Acid Dianhydride |
| CAS | 156028-26-1 |
| Molecular Formula | C24H4Cl4O6 |
| Quantity | 100 mg |
| Synonym | 5,6,12,13-tetrachloro-Perylo[3,4-cd:9,10-c'd']dipyran-1,3,8,10-tetrone, 5,6,12,13-Tetrachloroperylo[3,4-cd:9,10-c'd']dipyran-1,3,8,10-tetrone, 1,6,7,12-Tetrachloro-3,4:9,10-perylenetetracarboxylic dianhydride, 1,6,7,12-Tetrachloroperylene-3,4:9,10-tetracarboxylic acid dianhydride, 1,6,7,12-Tetrachloroperylene-3,4:9,10-tetracarboxylic dianhydride, 5,6,12,13-Tetrachloroperylenetetracarboxylic dianhydride |
| InChI Formula | InChI=1S/C24H4Cl4O6/c25-9-1-5-13-6(22(30)33-21(5)29)2-11(27)17-18-12(28)4-8-14-7(23(31)34-24(8)32)3-10(26)16(20(14)18)15(9)19(13)17/h1-4H |
| SMILES | C1=C2C3=C4C(=C1Cl)C5=C(C=C6C7=C5C(=C(C=C7C(=O)OC6=O)Cl)C4=C(C=C3C(=O)OC2=O)Cl)Cl |
| Molecular Weight (g/mol) | 530.1 |
| Formula Weight | 527.8762 |