Learn More
Alfa Aesar™ 1,5-Hexadien-3-ol, 95%
Product Code. 11312056
Change view
Click to view available options
Quantity:
2 g
10 g
Unit Size:
10g
2g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11312056 10 g 10g
11302056 2 g 2g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11312056 Supplier Alfa Aesar™ Supplier No. L03182.09
Request Bulk or Custom Format
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Applications
1,5-Hexadien-3-ol is used in the synthesis of (+)-obtusenyne. It is used as intermediates. It undergoes Sharpless kinetic resolution and ring-closing metathesis to yield nine membered oxocene.
Solubility
Sparingly Soluble.
Notes
Keep away from heat. Incompatible with Oxidizing agents.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 924-41-4
Molecular Formula C6H10O
Molecular Weight (g/mol) 98.145
MDL Number MFCD00008662
InChI Key SZYLTIUVWARXOO-UHFFFAOYSA-N
Synonym 1,5-hexadien-3-ol, 1,5-hexadiene-3-ol, 3-hydroxy-1,5-hexadiene
PubChem CID 79122
IUPAC Name hexa-1,5-dien-3-ol
SMILES C=CCC(C=C)O
Specifications
Density 0.877
Boiling Point 133°C to 134°C
Flash Point 29°C (84°F)
Refractive Index 1.448
Quantity 10 g
UN Number UN1987
Beilstein 1719962
Solubility Information Sparingly Soluble.
Formula Weight 98.15
Percent Purity 95%
Chemical Name or Material 1,5-Hexadien-3-ol