missing translation for 'onlineSavingsMsg'
Learn More

1,4-Naphthoquinone, Sigma-Aldrich™

Product Code. 15899290
Change view
Click to view available options
Quantity:
250 g
25 kg
Unit Size:
250g
25kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15899290 25 kg 25kg
15879290 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15899290 Supplier Sigma Aldrich Supplier No. 8062159025
 Restricted Item
Estimated Shipment: 26-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

For synthesis

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 130-15-4
Grade Research
Molecular Weight (g/mol) 158.15

Specifications

Melting Point 120°C to 125°C (Sublimed)
Density 1.42 g/cm3 at 20°C
MDL Number MFCD00001676
Quantity 25 kg
Synonym 1,4-Naphthoquinone, 1,4-Naphthalenedione, 1,4-Dihydro-1,4-diketonaphthalene, 1,4-Naphthoquinone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.