Learn More
1-(4-Methoxyphenyl)acetophenone, TRC
Specifications
Specifications
| Chemical Name or Material | 1-(4-Methoxyphenyl)acetophenone |
| CAS | 1023-17-2 |
| Molecular Formula | C15H14O2 |
| Quantity | 500 mg |
| Synonym | 1-(4-Methoxyphenyl)-2-phenyl-ethanone, 2-Phenyl-p-methoxyacetophenone, 4-Methoxy-alpha-phenylacetophenone, 4-Methoxydeoxybenzoin, Benzyl p-Methoxyphenyl Ketone, NSC 26658, p-Anisyl Benzyl Ketone |
| InChI Formula | InChI=1S/C15H14O2/c1-17-14-9-7-13(8-10-14)15(16)11-12-5-3-2-4-6-12/h2-10H,11H2,1H3 |
| SMILES | COc1ccc(cc1)C(=O)Cc2ccccc2 |
| IUPAC Name | 1-(4-methoxyphenyl)-2-phenylethanone |
| Molecular Weight (g/mol) | 226.27 |
| Formula Weight | 226.0994 |