missing translation for 'onlineSavingsMsg'
Learn More

1-(4-Fluorophenyl)vinylboronic acid pinacol ester, 97%, Thermo Scientific™

Product Code. 15499915
Change view
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15499915 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15499915 Supplier Thermo Scientific Alfa Aesar Supplier No. H52584.MD
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Specifications

CAS 850567-55-4
Melting Point 38°C to 42°C
Molecular Formula C14H18BFO2
MDL Number MFCD06659925
Quantity 250 mg
Synonym 1-4-fluorophenyl vinylboronic acid, pinacol ester, 2-1-4-fluorophenyl vinyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 2-1-4-fluorophenyl ethenyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 1-4-fluorophenyl vinylboronic acid pinacol ester, 1-4-fluorophenyl vinylboronic acid,pinacol ester, 4-fluoro-alpha-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl styrene
InChI Key BNROZDXKAQJDJC-UHFFFAOYSA-N
SMILES CC1(C)OB(OC1(C)C)C(=C)C1=CC=C(F)C=C1
IUPAC Name 2-[1-(4-fluorophenyl)ethenyl]-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
Molecular Weight (g/mol) 248.10
PubChem CID 44717770
Formula Weight 248.1
Percent Purity 97%
Chemical Name or Material 1-(4-Fluorophenyl)vinylboronic acid pinacol ester
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.