Learn More
1,4-Dioxane, anhydrous, 99.8%, stab. with 1-3ppm BHT, packaged under argon in resealable AcroSeal™ bottles
Product Code. 16388207
Click to view available options
Quantity:
100 mL
1 L
4 x 1 L
Unit Size:
1 L
100mL
4 L
Product Code. 16388207
Brand: Thermo Scientific Alfa Aesar 047305.M1
Request Bulk or Custom Format
CAS: 123-91-1 | C{4}H{8}O{2}
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Specifications
Melting Point 11.8°C
CAS 123-91-1
Boiling Point 100°C to 102°C
Quantity 4 x 1 L
Molecular Formula C{4}H{8}O{2}
Percent Purity 99.8%