Learn More
1,4-Dioxacycloheptadecane-5,17-dione, 98%, BLD Pharm™
Specifications
Specifications
| CAS | 105-95-3 |
| Molecular Formula | C15H26O4 |
| MDL Number | MFCD00046977 |
| Quantity | 100 g |
| SMILES | O=C(CCCCCCCCCCC1)OCCOC1=O |
| Molecular Weight (g/mol) | 270.36 |
| Percent Purity | 98% |
| Chemical Name or Material | 1,4-Dioxacycloheptadecane-5,17-dione |