Learn More
1,4-Dimethoxy-2-methylbenzene, TRC
Specifications
Specifications
| Chemical Name or Material | 1,4-Dimethoxy-2-methylbenzene |
| CAS | 24599-58-4 |
| Molecular Formula | C9H12O2 |
| Quantity | 2.5 g |
| Synonym | 2,5-Dimethoxytoluene |
| InChI Formula | InChI=1S/C9H12O2/c1-7-6-8(10-2)4-5-9(7)11-3/h4-6H,1-3H3 |
| SMILES | COc1ccc(OC)c(C)c1 |
| IUPAC Name | 1,4-dimethoxy-2-methylbenzene |
| Molecular Weight (g/mol) | 152.19 |
| Formula Weight | 152.0837 |