Learn More
1,4-Dibutyl Benzene-1,4-dicarboxylate, TRC
Product Code. 30011927
Change view
Click to view available options
Quantity:
100 mg
500 mg
1 g
Unit Size:
100mg
1g
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30011927 1 g 1g
30010942 500 mg 500mg
30007761 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30011927 Supplier Toronto Research Chemicals Supplier No. TRCD1593911G
 Restricted Item
Estimated Shipment: 27-08-2026
Add to basket
Add to basket
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 1962-75-0
Molecular Formula C16H22O4
Molecular Weight (g/mol) 278.34
Synonym 1,4-Benzenedicarboxylic acid, dibutyl ester (9CI), 1,4-Dibutyl 1,4-benzenedicarboxylate (ACI), Terephthalic acid, dibutyl ester (6CI, 7CI, 8CI), 1,4-Dibutyl benzene-1,4-dicarboxylate, Butyl terephthalate, Dibutyl terephthalate, Eastman DBT, NSC 6349, 1,4-Benzenedicarboxylic acid, 1,4-dibutyl ester (ACI), Terephthalic Acid, Bis-butyl Ester
IUPAC Name dibutyl benzene-1,4-dicarboxylate
SMILES CCCCOC(=O)c1ccc(cc1)C(=O)OCCCC
Specifications
Quantity 1 g
InChI Formula InChI=1S/C16H22O4/c1-3-5-11-19-15(17)13-7-9-14(10-8-13)16(18)20-12-6-4-2/h7-10H,3-6,11-12H2,1-2H3
Formula Weight 278.1518
Chemical Name or Material 1,4-Dibutyl Benzene-1,4-dicarboxylate