missing translation for 'onlineSavingsMsg'
Learn More

1,4-Diazabicyclo[2.2.2]octane, Sigma-Aldrich™

Product Code. 15899000
Change view
Click to view available options
Quantity:
100 g
250 g
1 kg
Unit Size:
100g
1kg
250g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15899000 100 g 100g
15809010 250 g 250g
15819010 1 kg 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15899000 Supplier Sigma-Aldrich Supplier No. 8034560100
 Restricted Item
Estimated Shipment: 23-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

For synthesis

We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product has been enhanced for catalytic efficiency.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 280-57-9
Grade Research
Molecular Weight (g/mol) 112.17

Specifications

Melting Point 155°C to 158°C
Density 1.14 g/cm3 at 25°C
Boiling Point 173.4°C/1000 hPa
MDL Number MFCD00006689
Quantity 100 g
Synonym 1,4-Diazabicyclo[2.2.2]octane, Triethylenediamine, DABCO, 1,4-Diazabicyclo[2.2.2]octane
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.