missing translation for 'onlineSavingsMsg'
Learn More

1,4-Diacetoxy-2-butyne, 97%, Thermo Scientific™

Product Code. 11423707
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11423707 25 g 25g
11413707 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11423707 Supplier Thermo Scientific Alfa Aesar Supplier No. A18182.14
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1573-17-7
Molecular Formula C8H10O4
Molecular Weight (g/mol) 170.16
MDL Number MFCD00014982
InChI Key TVIMIQVBDDNCCR-UHFFFAOYSA-N
Synonym 1,4-diacetoxy-2-butyne, but-2-yne-1,4-diyl diacetate, butynediol diacetate, 2-butyne-1,4-diol diacetate, 2-butyne-1,4-diol, diacetate, 2-butyne-1,4-diol, 1,4-diacetate, 4-acetyloxy but-2-yn-1-yl acetate, acmc-209dfx, 2-butyne-1, diacetate, bis acetoxymethyl acetylene
PubChem CID 74081
IUPAC Name 4-acetyloxybut-2-ynyl acetate
SMILES CC(=O)OCC#CCOC(C)=O

Specifications

Melting Point 27°C to 28°C
Density 1.125
Boiling Point 116°C to 118°C (8 mmHg)
Flash Point >110°C (230°F)
Linear Formula CH3CO2CH2C≡CCH2O2CCH3
Refractive Index 1.454
Quantity 25 g
Formula Weight 170.16
Percent Purity 97%
Chemical Name or Material 1,4-Diacetoxy-2-butyne
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.