missing translation for 'onlineSavingsMsg'
Learn More

1,4-Diacetoxy-2-butyne, 97%, Thermo Scientific™

Product Code. 11413707
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11413707 5 g 5g
11423707 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11413707 Supplier Thermo Scientific Alfa Aesar Supplier No. A18182.06

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemische Identifikatoren

CAS 1573-17-7
Molecular Formula C8H10O4
Molecular Weight (g/mol) 170.16
MDL Number MFCD00014982
InChI Key TVIMIQVBDDNCCR-UHFFFAOYSA-N
Synonym 1,4-diacetoxy-2-butyne, but-2-yne-1,4-diyl diacetate, butynediol diacetate, 2-butyne-1,4-diol diacetate, 2-butyne-1,4-diol, diacetate, 2-butyne-1,4-diol, 1,4-diacetate, 4-acetyloxy but-2-yn-1-yl acetate, acmc-209dfx, 2-butyne-1, diacetate, bis acetoxymethyl acetylene
PubChem CID 74081
IUPAC Name 4-acetyloxybut-2-ynyl acetate
SMILES CC(=O)OCC#CCOC(C)=O

Spezifikation

Melting Point 27°C to 28°C
Density 1.125
Boiling Point 116°C to 118°C (8 mmHg)
Flash Point >110°C (230°F)
Linear Formula CH3CO2CH2C≡CCH2O2CCH3
Refractive Index 1.454
Quantity 5 g
Formula Weight 170.16
Percent Purity 97%
Chemical Name or Material 1,4-Diacetoxy-2-butyne
Name des Produkts
Wählen Sie ein Thema

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.