Learn More
1,4-Di(hydroxymethyl)benzene, TRC
Specifications
Specifications
| CAS | 589-29-7 |
| Molecular Formula | C8 H10 O2 |
| Quantity | 1 g |
| Synonym | 1,4-Benzenedimethanol, p-Xylene-α,α'-diol (6CI,7CI,8CI), 1,4-Bis(hydroxymethyl)benzene, 1,4-Di(hydroxymethyl)benzene, 1,4-Dimethylolbenzene, 1,4-Phenylenedimethanol, 1,4-Xylylene glycol, 4-(Hydroxymethyl)benzenemethanol, 4-(Hydroxymethyl)benzyl alcohol, NSC 5097, Terephthalyl alcohol, [4-(Hydroxymethyl)phenyl]methanol, p-(Hydroxymethyl)benzyl alcohol, p-Benzenedimethanol, p-Bis(hydroxymethyl)benzene, p-Xylene dimethanol, p-Xylene glycol, p-Xylylene glycol, p-Xylylene-α,α'-diol, p-Xylylenediol, α,α'-Dihydroxy-p-xylene, α,α'-p-Xylenediol |
| InChI Formula | InChI=1S/C8H10O2/c9-5-7-1-2-8(6-10)4-3-7/h1-4,9-10H,5-6H2 |
| SMILES | OCc1ccc(CO)cc1 |
| IUPAC Name | [4-(hydroxymethyl)phenyl]methanol |
| Molecular Weight (g/mol) | 138.16 |
| Formula Weight | 138.0681 |
| Chemical Name or Material | 1,4-Phenylenedimethanol |