Learn More
1-[4-Chloro-2-(2-fluorobenzoyl)phenyl]-2-methyl-1H-imidazole-5-carboxaldehyde, Mikromol™
Description
1-[4-Chloro-2-(2-fluorobenzoyl)phenyl]-2-methyl-1H-imidazole-5-carboxaldehyde
Specifications
Specifications
| Chemical Name or Material | 1-[4-Chloro-2-(2-fluorobenzoyl)phenyl]-2-methyl-1H-imidazole-5-carboxaldehyde |
| CAS | 2454678-38-5 |
| Analyte or Component Names | 1-[4-Chloro-2-(2-fluorobenzoyl)phenyl]-2-methyl-1H-imidazole-5-carboxaldehyde |
| Shipping Condition | Room Temperature |
| Molecular Formula | C18 H12 Cl F N2 O2 |
| Certifications/Compliance | ISO 9001 |
| Synonym | 1-(4-Chloro-2-(2-fluorobenzoyl)phenyl)-2-methyl-1H-imidazole-5-carboxaldehyde, 3-[4-Chloro-2-(2-fluorobenzoyl)phenyl]-2-methyl-imidazole-4-carbaldehyde |
| InChI Formula | InChI=1S/C18H12ClFN2O2/c1-11-21-9-13(10-23)22(11)17-7-6-12(19)8-15(17)18(24)14-4-2-3-5-16(14)20/h2-10H,1H3 |
| SMILES | Cc1ncc(C=O)n1c2ccc(Cl)cc2C(=O)c3ccccc3F |
| IUPAC Name | 3-[4-chloro-2-(2-fluorobenzoyl)phenyl]-2-methyl-imidazole-4-carbaldehyde |
| Show More |