missing translation for 'onlineSavingsMsg'
Learn More

1,4-Bis(trans-2-phenylethenyl)benzene, 97%, Thermo Scientific™

Product Code. 15454087
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15454087 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15454087 Supplier Thermo Scientific Alfa Aesar Supplier No. H31436.03

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Specifications

CAS 1608-41-9
Melting Point 264°C to 266°C
Molecular Formula C22H18
MDL Number MFCD00154950
Quantity 1 g
Synonym 1,4-distyrylbenzene, 1,4-bis e-2-phenylethenyl benzene, trans,trans-1,4-distyrylbenzene, 1,4-bis trans-2-phenylethenyl benzene, benzene, 1,4-bis 2-phenylethenyl, benzene, 1,4-bis 1e-2-phenylethenyl, 1,4-bis trans-styryl benzene, 1,4-bis e-styryl benzene, 4-e-styryl-cis-stilbene, 1,4-di trans-beta-styryl benzene
InChI Key IJAAWBHHXIWAHM-PHEQNACWSA-N
SMILES C1=CC=C(C=C1)C=CC2=CC=C(C=C2)C=CC3=CC=CC=C3
IUPAC Name 1,4-bis[(E)-2-phenylethenyl]benzene
Molecular Weight (g/mol) 282.386
PubChem CID 640299
Formula Weight 282.39
Percent Purity 97%
Chemical Name or Material 1,4-Bis(trans-2-phenylethenyl)benzene
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.