Learn More
1,4-Bis(aminomethyl)-benzene, Sigma-Aldrich™
Specifications
Specifications
| CAS | 539-48-0 |
| Melting Point | 60°C to 64°C |
| Boiling Point | 230°C/13.33 hPa |
| MDL Number | MFCD00009821 |
| Quantity | 10 g |
| Synonym | 1,4-Bis(aminomethyl)-benzene, 1,4-Bis(aminomethyl)-benzene |
| Molecular Weight (g/mol) | 136.19 |
| Grade | Research |