missing translation for 'onlineSavingsMsg'
Learn More

1,4-Bis(2-hydroxyisopropyl)benzene, 97%

Product Code. 11457004
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11457004 5 g 5g
11447004 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11457004 Supplier Thermo Scientific Alfa Aesar Supplier No. B21472.06
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 2948-46-1 | C12H18O2 | 194.274 g/mol

1,4-Bis(2-hydroxyisopropyl)benzene is used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,4-Bis(2-hydroxyisopropyl)benzene is used as a pharmaceutical intermediate.

Solubility
Slightly Soluble in water (2.1 g/L) (25°C).

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from acids, acid chlorides, acid anhydrides, strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2948-46-1
Molecular Formula C12H18O2
Molecular Weight (g/mol) 194.274
MDL Number MFCD00009827
InChI Key LEARFTRDZQQTDN-UHFFFAOYSA-N
Synonym 1,4-bis 2-hydroxy-2-propyl benzene, 1,4-bis 1-methyl-1-hydroxyethyl benzene, alpha,alpha,alpha',alpha'-tetramethyl-1,4-benzenedimethanol, unii-g785drb5xh, p-bis 2-hydroxyisopropyl benzene, 1,4-bis 2-hydroxyisopropyl benzene, p-bis alpha-hydroxyisopropyl benzene, g785drb5xh, alpha,alpha'-dihydroxy-p-diisopropylbenzene, alpha,alpha,alpha',alpha'-tetramethyl-p-xylylenediol
PubChem CID 18062
IUPAC Name 2-[4-(2-hydroxypropan-2-yl)phenyl]propan-2-ol
SMILES CC(C)(C1=CC=C(C=C1)C(C)(C)O)O

Specifications

Melting Point 142°C to 146°C
Boiling Point 167°C to 168°C (12 mmHg)
Quantity 5 g
Solubility Information Slightly Soluble in water (2.1g/L) (25°C).
Formula Weight 194.28
Percent Purity 97%
Chemical Name or Material 1,4-Bis(2-hydroxyisopropyl)benzene

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.