Learn More
1,4,7-Trimethyl-1,4,7-triazacyclononane, TRC
Specifications
Specifications
| Chemical Name or Material | 1,4,7-Trimethyl-1,4,7-triazacyclononane |
| CAS | 96556-05-7 |
| Molecular Formula | C9H21N3 |
| Quantity | 500 mg |
| Synonym | Octahydro-1,4,7-trimethyl-1H-1,4,7-triazonine, 1,4,7-Trimethyl-1,4,7-triazonane, N,N',N''-Trimethyl-1,4,7-triazacyclononane, TMTAN |
| InChI Formula | InChI=1S/C9H21N3/c1-10-4-6-11(2)8-9-12(3)7-5-10/h4-9H2,1-3H3 |
| SMILES | CN1CCN(C)CCN(C)CC1 |
| IUPAC Name | 1,4,7-trimethyl-1,4,7-triazonane |
| Molecular Weight (g/mol) | 171.28 |
| Formula Weight | 171.1735 |
Product Title
Vous avez repéré une opportunité d'amélioration ?