missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 1,4,7,10-Tetrakis(ethoxycarbonylmethyl)-1,4,7,10-tetraazacyclododecane

Product Code. 15457605
Change view
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15457605 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15457605 Supplier Alfa Aesar™ Supplier No. H26646.MD
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

CAS 137076-50-7
Molecular Formula C24H44N4O8
MDL Number MFCD09263313
Quantity 250 mg
Sensitivity Hygroscopic
Synonym dotaet, 1,4,7,10-tetrakis ethoxycarbonylmethyl-1,4,7,10-tetraazacyclododecane, acmc-20ap3l, cyclen-n,n',n',n-tetraacetic acid tetraethyl ester, 1,4,7,10-tetrakis ethoxycarbonylmethyl-1,4,7,10-tetraazacyc, 1,4,7,10-tetraazacyclododecane-1,4,7,10-tetraacetic acid tetraethyl ester, ethyl 2-4,7,10-tris 2-ethoxy-2-oxoethyl-1,4,7,10-tetrazacyclododec-1-yl acetate, tetraethyl 2,2',2,2'-1,4,7,10-tetraazacyclododecane-1,4,7,10-tetrayl tetraacetate, ethyl 2-4,7,10-tris 2-ethoxy-2-oxoethyl-1,4,7,10-tetraazacyclododecan-1-yl acetate
InChI Key SHXTUKRRXWXLMI-UHFFFAOYSA-N
SMILES CCOC(=O)CN1CCN(CC(=O)OCC)CCN(CC(=O)OCC)CCN(CC(=O)OCC)CC1
IUPAC Name ethyl 2-[4,7,10-tris(2-ethoxy-2-oxoethyl)-1,4,7,10-tetrazacyclododec-1-yl]acetate
Molecular Weight (g/mol) 516.64
PubChem CID 5255369
Formula Weight 516.64
Chemical Name or Material 1,4,7,10-Tetrakis(ethoxycarbonylmethyl)-1,4,7,10-tetraazacyclododecane
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.