missing translation for 'onlineSavingsMsg'
Learn More

1-[4-(1,1-Dimethylethyl)phenyl]-4-[4-(hydroxydiphenylmethyl)piperidin-1-yl]butan-1-one (Terfenadone), Mikromol™

Product Code. 30464224
Change view
Click to view available options
Quantity:
25 mg
100 mg
4 x 25 mg
Unit Size:
100mg
25mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30464224 4 x 25 mg -
30050025 25 mg 25mg
17917244 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30464224 Supplier Mikromol Supplier No. MM0055.0400254AMP
 Restricted Item
Estimated Shipment: 17-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

1-[4-(1,1-Dimethylethyl)phenyl]-4-[4-(hydroxydiphenylmethyl)piperidin-1-yl]butan-1-one (Terfenadone), Mikromol
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Terfenadone
CAS 43076-30-8
Analyte or Component Names Terfenadone
Shipping Condition Room Temperature
Molecular Formula C32 H39 N O2
Certifications/Compliance ISO 9001
Synonym 1-[4-(1,1-Dimethylethyl)phenyl]-4-[4-(hydroxydiphenylmethyl)piperidin-1-yl]butan-1-one, Terfenadine Imp. A (EP), Terfenadone, Terfenadine Related Compound A, Terfenadine Impurity A
InChI Formula InChI=1S/C32H39NO2/c1-31(2,3)26-18-16-25(17-19-26)30(34)15-10-22-33-23-20-29(21-24-33)32(35,27-11-6-4-7-12-27)28-13-8-5-9-14-28/h4-9,11-14,16-19,29,35H,10,15,20-24H2,1-3H3
Solution Type Neat
SMILES CC(C)(C)c1ccc(cc1)C(=O)CCCN2CCC(CC2)C(O)(c3ccccc3)c4ccccc4
IUPAC Name 1-(4-tert-butylphenyl)-4-[4-[hydroxy(diphenyl)methyl]piperidin-1-yl]butan-1-one
Molecular Weight (g/mol) 469.66
Quantity 4 x 25 mg
Formula Weight 469.2981 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.