missing translation for 'onlineSavingsMsg'
Learn More

1,3-Phenylenediamine, Sigma-Aldrich™

Product Code. 15891311
Change view
Click to view available options
Quantity:
5 g
250 g
1 kg
Unit Size:
1kg
250g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15891311 5 g 5g
15801321 250 g 250g
15811321 1 kg 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15891311 Supplier Sigma Aldrich Supplier No. 8209910005
 Restricted Item
Estimated Shipment: 16-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

For synthesis

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 108-45-2
Grade Research
Molecular Formula 1,3-(NH2)2C6H4
Molecular Weight (g/mol) 108.14

Specifications

Melting Point 63°C to 64°C
Density 1.14 g/cm3 at 20°C
Boiling Point 283°C to 284°C/1013 hPa
MDL Number MFCD00007799
Quantity 5 g
Synonym 1,3-Phenylenediamine, 1,3-Phenylenediamine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.