Learn More
1,3-Indanedione, Sigma-Aldrich™
Product Code. 15841121
Change view
Click to view available options
Quantity:
10 g
Unit Size:
10g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15841121 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
MQ200 Graded - For Research and Non-regulated Industrial Applications
Product Code. 15841121 Supplier Sigma Aldrich Supplier No. 8206980010
 Restricted Item
Estimated Shipment: 26-11-2026
MQ200 Graded - For Research and Non-regulated Industrial Applications
For synthesis
TRUSTED_SUSTAINABILITY
Specifications
CAS 606-23-5
Melting Point 129°C to 132°C
MDL Number MFCD00003779
Quantity 10 g
Synonym 1,3-Indanedione, 1,3-Hydrindenedione, 1,3-Dioxoindane, 1,3-Indanedione
Molecular Weight (g/mol) 146.14
Grade Research