missing translation for 'onlineSavingsMsg'
Learn More

1,3-Dioxolane, Sigma-Aldrich™

Product Code. 15829050
Change view
Click to view available options
Quantity:
100 mL
500 mL
Unit Size:
100mL
500mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15829050 500 mL 500mL
15819050 100 mL 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15829050 Supplier Sigma Aldrich Supplier No. 8035530500
 Restricted Item
Estimated Shipment: 28-01-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Stabilized for synthesis

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 646-06-0
Grade Research
Molecular Weight (g/mol) 74.08

Specifications

Melting Point -95°C
Density 1.065 g/cm3 at 20°C
Boiling Point 74°C to 75°C/1013 hPa
MDL Number MFCD00003207
Quantity 500 mL
Synonym 1,3-Dioxolane, 1,3-Dioxacyclopentane, Formaldehyde ethylene acetal, Ethylene glycol methylene ether, 1,3-Dioxolane
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.