Learn More
1,3-Dimethyl-5-pyrazolone, TRC
Specifications
Specifications
| CAS | 2749-59-9 |
| Molecular Formula | C5H8N2O |
| Quantity | 10 g |
| Synonym | 2,4-Dihydro-2,5-dimethyl-3H-pyrazol-3-one, 1,3-Dimethyl-5-pyrazolinone, 1,3-Dimethyl-5-pyrazolone, NSC 304 |
| InChI Formula | InChI=1S/C5H8N2O/c1-4-3-5(8)7(2)6-4/h3H2,1-2H3 |
| SMILES | CN1N=C(C)CC1=O |
| Molecular Weight (g/mol) | 112.13 |
| Formula Weight | 112.0637 |
| Chemical Name or Material | 1,3-Dimethyl-5-pyrazolone |