Learn More
1,3-Diiodobenzene, TRC
Specifications
Specifications
| Chemical Name or Material | 1,3-Diiodobenzene |
| CAS | 626-00-6 |
| Molecular Formula | C6H4I2 |
| Quantity | 25 g |
| Synonym | m-Diiodobenzene |
| InChI Formula | InChI=1S/C6H4I2/c7-5-2-1-3-6(8)4-5/h1-4H |
| SMILES | IC1=CC=CC(I)=C1 |
| IUPAC Name | 1,3-diiodobenzene |
| Molecular Weight (g/mol) | 329.9 |
| Formula Weight | 329.84 |