missing translation for 'onlineSavingsMsg'
Learn More

1,3-Dibromo-5-chlorobenzene, 98%, Thermo Scientific Chemicals

Product Code. p-7028221
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11431030 1 g 1g
11441030 5 g 5g
11451030 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11431030 Supplier Thermo Scientific Alfa Aesar Supplier No. B25411.03

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 14862-52-3 | Molecular Formula: C6H3Br2Cl | Molecular Weight: 270.348 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14862-52-3
Molecular Formula C6H3Br2Cl
Molecular Weight (g/mol) 270.348
MDL Number MFCD00070765
InChI Key FNKCOUREFBNNHG-UHFFFAOYSA-N
Synonym 3,5-dibromochlorobenzene, 1-chloro-3,5-dibromobenzene, benzene, 1,3-dibromo-5-chloro, pubchem3634, ambsclk-224, intermediates-zcf02139, ksc181m3d, 1,3-dibromo-5-chloro-benzene, tpc-i027, fnkcourefbnnhg-uhfffaoysa
PubChem CID 84676
IUPAC Name 1,3-dibromo-5-chlorobenzene
SMILES C1=C(C=C(C=C1Br)Br)Cl

Specifications

Melting Point 95°C to 98°C
Quantity 1 g
Formula Weight 270.35
Percent Purity 98%
Chemical Name or Material 1,3-Dibromo-5-chlorobenzene

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.