missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,3-Diaminopropane, Sigma-Aldrich™
Specifica
Specifica
| Melting Point | -12°C |
| Density | 0.89 g/cm3 at 20°C |
| Boiling Point | 137°C to 140°C/1013 mbar |
| MDL Number | MFCD00008228 |
| Quantity | 500 mL |
| Synonym | 1,3-Diaminopropane, 1,3-Propanediamine, Trimethylenediamine, 1,3-Diaminopropane |
Titolo del prodotto
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?