Learn More
1,3-Diamino-2-propanol, TRC
Product Code. 30363187
Change view
Click to view available options
Quantity:
10 g
25 g
100 g
Unit Size:
100g
10g
25g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30363187 25 g 25g
30359926 10 g 10g
30362445 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30363187 Supplier Toronto Research Chemicals Supplier No. TRCD41663025G
 Restricted Item
Estimated Shipment: 24-11-2026
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 616-29-5
Molecular Formula C3H10N2O
Molecular Weight (g/mol) 90.12
Synonym (3-Amino-2-hydroxypropyl)amine, 1,3-Diamino-2-hydroxypropane, 1,3-Diamino-2-propanol, 1,3-Diaminopropan-2-ol, 2-Hydroxy-1,3-diaminopropane, 2-Hydroxy-1,3-propanediamine, 2-Hydroxypropylenediamine, Dapol, NSC 6070
IUPAC Name 1,3-diaminopropan-2-ol
SMILES NCC(O)CN
Specifications
Quantity 25 g
InChI Formula InChI=1S/C3H10N2O/c4-1-3(6)2-5/h3,6H,1-2,4-5H2
Formula Weight 90.0793
Chemical Name or Material 1,3-Diamino-2-propanol