missing translation for 'onlineSavingsMsg'
Learn More

1,3-Diamino-2-propanol, 97%

Product Code. p-7024623
Change view
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11454167 10 g 10g
11464167 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11454167 Supplier Thermo Scientific Alfa Aesar Supplier No. A19618.09
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 616-29-5 | C3H10N2O | 90.126 g/mol

It finds its application as the formaldehyde absorbent, carbon dioxide and acid gas absorption. It is also used as a pharmaceutical intermediate and in organic synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It finds its application as the formaldehyde absorbent, carbon dioxide and acid gas absorption. It is also used as a pharmaceutical intermediate and in organic synthesis.

Solubility
Very soluble in water.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 616-29-5
Molecular Formula C3H10N2O
Molecular Weight (g/mol) 90.126
MDL Number MFCD00008142
InChI Key UYBWIEGTWASWSR-UHFFFAOYSA-N
Synonym 1,3-diamino-2-propanol, 1,3-diamino-2-hydroxypropane, diaminoisopropanol, 2-hydroxy-1,3-diaminopropane, 2-propanol, 1,3-diamino, 2-hydroxy-1,3-propanediamine, 1,3-diamino-2-hydroxy-propane, 2-propanol,3-diamino, acmc-1b1xv, 1,3 diamino-propan-2-ol
PubChem CID 61157
IUPAC Name 1,3-diaminopropan-2-ol
SMILES C(C(CN)O)N

Specifications

Melting Point 40°C to 45°C
Boiling Point 235°C
Flash Point >110°C (230°F)
Quantity 10 g
Beilstein 741859
Solubility Information Very soluble in water.
Formula Weight 90.13
Percent Purity 97%
Chemical Name or Material 1,3-Diamino-2-propanol
compliance-icons
Product Identifier
  • 1,3-Diamino-2-propanol
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
  • MIXTURE LIST-Contain: 1,3-Diamino-2-propanol

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.