missing translation for 'onlineSavingsMsg'
Learn More

1,3-Di-HABA Kanamycin A, TRC

Product Code. 30011067
Change view
Click to view available options
Quantity:
1 mg
10 mg
25 mg
Unit Size:
10mg
1mg
25mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30011067 1 mg 1mg
30137841 25 mg 25mg
30012586 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30011067 Supplier Toronto Research Chemicals Supplier No. TRCD4463751MG
 Restricted Item
Estimated Shipment: 06-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 927821-99-6
Molecular Formula C26 H50 N6 O15
Molecular Weight (g/mol) 686.71
Synonym D-Streptamine, O-3-amino-3-deoxy-α-D-glucopyranosyl-(1→6)-O-[6-amino-6-deoxy-α-D-glucopyranosyl-(1→4)]-N1,N3-bis[(2S)-4-amino-2-hydroxy-1-oxobutyl]-2-deoxy-, O-3-Amino-3-deoxy-α-D-glucopyranosyl-(1→6)-O-[6-amino-6-deoxy-α-D-glucopyranosyl-(1→4)]-N1,N3-bis[(2S)-4-amino-2-hydroxy-1-oxobutyl]-2-deoxy-D-streptamine, 4-O-(3-Amino-3-deoxy-α-D-glucopyranosyl)-6-O-(6-amino-6-deoxy-α-D-glucopyranosyl)-1,3-N-bis[(2S)-4-amino-2-hydroxybutanoyl]-2-deoxy-L-streptamine, 1,3-Di-HABA Kanamycin A
IUPAC Name (2S)-4-amino-N-[(1R,2S,3R,4R,5S)-2-[(2S,3R,4S,5S,6R)-4-amino-3,5-dihydroxy-6-(hydroxymethyl)oxan-2-yl]oxy-5-[[(2S)-4-amino-2-hydroxybutanoyl]amino]-4-[(2R,3R,4S,5S,6R)-6-(aminomethyl)-3,4,5-trihydroxyoxan-2-yl]oxy-3-hydroxycyclohexyl]-2-hydroxybutanamide
SMILES NCC[C@H](O)C(=O)N[C@H]1C[C@@H](NC(=O)[C@@H](O)CCN)[C@H](O[C@H]2O[C@H](CO)[C@@H](O)[C@H](N)[C@H]2O)[C@@H](O)[C@@H]1O[C@H]3O[C@H](CN)[C@@H](O)[C@H](O)[C@H]3O

Specifications

Quantity 1 mg
InChI Formula InChI=1S/C26H50N6O15/c27-3-1-10(34)23(42)31-8-5-9(32-24(43)11(35)2-4-28)22(47-26-19(40)18(39)16(37)12(6-29)44-26)20(41)21(8)46-25-17(38)14(30)15(36)13(7-33)45-25/h8-22,25-26,33-41H,1-7,27-30H2,(H,31,42)(H,32,43)/t8-,9+,10+,11+,12-,13-,14+,15-,16-,17-,18+,19-,20-,21+,22-,25-,26-/m1/s1
Formula Weight 686.3334
Percent Purity >95%
Chemical Name or Material 1,3-Di-HABA Kanamycin A
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.