Learn More
1,3,6-Tribromo-9H-carbazole, TRC
Product Code. 30456592
Change view
Click to view available options
Quantity:
5 mg
25 mg
50 mg
Unit Size:
25mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30456592 50 mg -
30455817 5 mg -
30455980 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30456592 Supplier Toronto Research Chemicals Supplier No. TRCT79827550MG
 Restricted Item
Estimated Shipment: 12-11-2026
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 55119-10-3
Molecular Formula C12H6Br3N
Molecular Weight (g/mol) 403.89
Synonym 1,3,6-Tribromocarbazole, NSC 121207
IUPAC Name 1,3,6-tribromo-9H-carbazole
SMILES Brc1ccc2[nH]c3c(Br)cc(Br)cc3c2c1
Specifications
Quantity 50 mg
InChI Formula InChI=1S/C12H6Br3N/c13-6-1-2-11-8(3-6)9-4-7(14)5-10(15)12(9)16-11/h1-5,16H
Formula Weight 400.805
Chemical Name or Material 1,3,6-Tribromo-9H-carbazole