missing translation for 'onlineSavingsMsg'
Learn More

1,3,6-Tribromo-9H-carbazole, TRC

Product Code. 30455817
Change view
Click to view available options
Quantity:
5 mg
25 mg
50 mg
Unit Size:
25mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30455817 5 mg -
30456592 50 mg -
30455980 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30455817 Supplier Toronto Research Chemicals Supplier No. TRCT7982755MG
 Restricted Item
Estimated Shipment: 12-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 55119-10-3
Molecular Formula C12H6Br3N
Molecular Weight (g/mol) 403.89
Synonym 1,3,6-Tribromocarbazole, NSC 121207
IUPAC Name 1,3,6-tribromo-9H-carbazole
SMILES Brc1ccc2[nH]c3c(Br)cc(Br)cc3c2c1

Specifications

Quantity 5 mg
InChI Formula InChI=1S/C12H6Br3N/c13-6-1-2-11-8(3-6)9-4-7(14)5-10(15)12(9)16-11/h1-5,16H
Formula Weight 400.805
Chemical Name or Material 1,3,6-Tribromo-9H-carbazole
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.