missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,3,5-Tris(3,5-di-tert-butyl-4-hydroxybenzyl)-2,4,6-trimethylbenzene, Mikromol
Description
1,3,5-Tris(3,5-di-tert-butyl-4-hydroxybenzyl)-2,4,6-trimethylbenzene, Mikromol
Specifications
Specifications
| Chemical Name or Material | Plastic additive 10 |
| CAS | 1709-70-2 |
| Analyte or Component Names | Plastic additive 10 |
| Shipping Condition | Room Temperature |
| Molecular Formula | C54 H78 O3 |
| Certifications/Compliance | ISO 9001 |
| Synonym | 4,4',4''-[(2,4,6-Trimethyl-1,3,5-benzenetriyl)tris(methylene)]tris[2,6-bis(1,1-dimethylethyl)phenol, α,α',α''-(2,4,6-Trimethyl-s-phenenyl)tris[2,6-di-tert-butyl-p-cresol, α,α',α''-(Trimethyl-s-phenenyl)tris[2,6-di-tert-butyl-p-cresol, 1,3,5-Dimethyl-2,4,6-tris(3,5-tert-butyl-4-hydroxybenzyl)benzene, 1,3,5-Trimethyl-2,4,6-tri(3,5-di-tert-butyl-4-hydroxybenzyl)benzene, 1,3,5-Trimethyl-2,4,6-tris(3,5-di-tert-butyl-4-hydroxybenzyl)benzene, 1,3,5-Trimethyl-2,4,6-tris(3,5-di-tert-butyl-4-hydroxytolyl)benzene, 1,3,5-Trimethyl-2,4,6-tris(3',5'-di-tert-butyl-4'-hydroxybenzyl)benzene, 1,3,5-Trimethyl-2,4,6-tris(4-hydroxy-3,5-di-tert-butylbenzyl)benzene, 1,3,5-Tris(3,5-di-tert-butyl-4-hydroxybenzyl)-2,4,6-trimethylbenzene, 1,3,5-Tris(3,5-di-tert-butyl-4-hydroxyphenylmethyl)-2,4,6-trimethylbenzene, 1,3,5-Trismethyl-2,4,6-tris(3,5-tert-butyl-4-hydroxybenzyl)benzene, 2,4,6-Bis(4-hydroxy-3,5-di-tert-butylbenzyl)mesitylene, 2,4,6-Tris(3,5-di-tert-butyl-4-hydroxybenzyl)mesitylene, 2,4,6-Tris(3',5'-di-tert-butyl-4'-hydroxybenzyl)mesitylene, 2,4,6-Tris(4-hydroxy-3,5-di-tert-butylbenzyl)-1,3,5-trimethylbenzene, 2,4,6-Tris(4-hydroxy-3,5-di-tert-butylbenzyl)mesitylene, 3,3',3'',5,5',5''-Hexa-tert-butyl-α,α',α''-(mesitylene-2,4,6-triyl)tri-p-cresol, ADK Stab A 330, ADK Stab AO 330, AO 330, AO 40, Agidol 40, Alvinox 100, Anox 330, Anox 330TDS, Antioxidant 1330, Antioxidant 330, Antioxidant 40, Ethanox 300, Ethanox 330, Ethyl 330, Ethyl Antioxidant 330, Good-rite 1330, IR 1330, Ionox 330, Irg 1330, Irganox 1330, Irganox 330, Mark AO 330, NSC 85846, Seenox 326M, Yoshinox 1330, Plastic additive 10 |
| InChI Formula | InChI=1S/C54H78O3/c1-31-37(22-34-25-40(49(4,5)6)46(55)41(26-34)50(7,8)9)32(2)39(24-36-29-44(53(16,17)18)48(57)45(30-36)54(19,20)21)33(3)38(31)23-35-27-42(51(10,11)12)47(56)43(28-35)52(13,14)15/h25-30,55-57H,22-24H2,1-21H3 |
| Solution Type | Neat |
| SMILES | Cc1c(Cc2cc(c(O)c(c2)C(C)(C)C)C(C)(C)C)c(C)c(Cc3cc(c(O)c(c3)C(C)(C)C)C(C)(C)C)c(C)c1Cc4cc(c(O)c(c4)C(C)(C)C)C(C)(C)C |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?