Learn More
1,3,5-Tribromo-2,4,6-Trimethylbenzene, TRC
Product Code. 30455810
Change view
Click to view available options
Quantity:
100 mg
500 mg
1 g
Unit Size:
100mg
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30455810 1 g -
30454365 100 mg 100mg
30455100 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30455810 Supplier Toronto Research Chemicals Supplier No. TRCT7983281G
 Restricted Item
Estimated Shipment: 11-11-2026
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 608-72-0
Molecular Formula C9H9Br3
Molecular Weight (g/mol) 356.88
IUPAC Name 1,3,5-tribromo-2,4,6-trimethylbenzene
SMILES Cc1c(Br)c(C)c(Br)c(C)c1Br
Specifications
Quantity 1 g
InChI Formula InChI=1S/C9H9Br3/c1-4-7(10)5(2)9(12)6(3)8(4)11/h1-3H3
Formula Weight 353.8254
Chemical Name or Material 1,3,5-Tribromo-2,4,6-Trimethylbenzene