Learn More
1,2-Naphthalic Anhydride, TRC
Specifications
Specifications
| Chemical Name or Material | 1,2-Naphthalic Anhydride |
| CAS | 5343-99-7 |
| Molecular Formula | C12H6O3 |
| Quantity | 50 mg |
| InChI Formula | InChI=1S/C12H6O3/c13-11-9-6-5-7-3-1-2-4-8(7)10(9)12(14)15-11/h1-6H |
| SMILES | O=C1OC(=O)c2c1ccc3ccccc23 |
| IUPAC Name | benzo[e][2]benzofuran-1,3-dione |
| Molecular Weight (g/mol) | 198.17 |
| Formula Weight | 198.0317 |