missing translation for 'onlineSavingsMsg'
Learn More

1-(2-Hydroxyethyl)-2-hydroxymethyl-5-nitroimidazole (Hydroxymetronidazole), Mikromol™

Product Code. 30049777
Change view
Click to view available options
Quantity:
25 mg
100 mg
Unit Size:
100mg
25mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30049777 25 mg 25mg
17972186 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30049777 Supplier Mikromol Supplier No. MM0012.040025
 Restricted Item
Estimated Shipment: 17-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

1-(2-Hydroxyethyl)-2-hydroxymethyl-5-nitroimidazole (Hydroxymetronidazole)
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Metronidazole-hydroxy
CAS 4812-40-2
Analyte or Component Names Metronidazole-hydroxy
Shipping Condition Room Temperature
Molecular Formula C6 H9 N3 O4
Certifications/Compliance ISO 17025
Synonym 1-(2-Hydroxyethyl)-2-hydroxymethyl-5-nitroimidazole, Hydroxymetronidazole, 2-(Hydroxymethyl)-5-nitroimidazole-1-ethanol
InChI Formula InChI=1S/C6H9N3O4/c10-2-1-8-5(4-11)7-3-6(8)9(12)13/h3,10-11H,1-2,4H2
SMILES OCCn1c(CO)ncc1[N+](=O)[O-]
IUPAC Name 2-[2-(hydroxymethyl)-5-nitroimidazol-1-yl]ethanol
Molecular Weight (g/mol) 187.15
Quantity 25 mg
Formula Weight 187.0593 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.