Learn More
1,2-Distearoyl-sn-glycero-3-phosphorylethanolamine, MedChemExpress
Product Code. 16495476
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16495476 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 16495476 Supplier Medchem Express Supplier No. HY112530
 Restricted Item
Estimated Shipment: 03-11-2026
MedChemExpress 1,2-Distearoyl-sn-glycero-3-phosphorylethanolamine (DSPE) is a phosphoethanolamine (PE) lipid that can be used in the synthesis of liposomes.
1,2-Distearoyl-sn-glycero-3-phosphorylethanolamine (DSPE) is a phosphoethanolamine (PE) lipid that can be used in the synthesis of liposomes.
Specifications
Chemical Name or Material 1,2-Distearoyl-sn-glycero-3-phosphorylethanolamine
CAS 1069-79-0
Color White
Purity Grade Notes Research
Molecular Formula C41H82NO8P
Quantity 100 mg
Synonym DSPE
Solubility Information H2O : 1 mg/mL (1.34 mM; Need ultrasonic) ∣Ethanol : < 1 mg/mL (ultrasonic;adjust pH to 5 with HCl) (insoluble) ∣DMSO : < 1 mg/mL (ultrasonic) (insoluble or slightly soluble)
SMILES CCCCCCCCCCCCCCCCCC(O[C@@H](COP(OCCN)(O)=O)COC(CCCCCCCCCCCCCCCCC)=O)=O
Molecular Weight (g/mol) 748.07
Formula Weight 748.07
Percent Purity 97.0%
Grade Research
Physical Form Solid
For Use With (Application) Metabolism-sugar/lipid metabolism