Learn More
1,2-Diphenylethane-d14, CDN
Product Code. 30315835
Change view
Click to view available options
Quantity:
0.5 g
1 g
Unit Size:
0.5g
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30315835 1 g 1g
30315091 0.5 g 0.5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30315835 Supplier CDN Supplier No. CDND12331G
 Restricted Item
Estimated Shipment: 13-08-2026
Add to basket
Add to basket
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material 1,2-Diphenylethane-D14
CAS 94371-89-8
Molecular Formula C14 D14
Quantity 1 g
Synonym Benzene-d5, 6,6'-(1,2-ethanediyl-1,1,2,2-d4)bis- (9 CI), Bibenzyl-d14 ̧ 6,6'-(1,2-Ethanediyl-1,1,2,2-d4)bis[benzene-d5], 1,2-Diphenylethane-d14, 1,2-Diphenylethane-D14
InChI Formula InChI=1 S/C14H14/c1-3-7-13(8-4-1)11-12-14-9-5-2-6-10-14/h1-10 H,11-12H2/i1D,2 D,3 D,4 D,5 D,6 D,7 D,8 D,9 D,10 D,11D2,12D2
SMILES [2 H]c1c([2 H])c([2 H])c(c([2 H])c1[2 H])C([2 H])([2 H])C([2 H])([2 H])c2c([2 H])c([2 H])c([2 H])c([2 H])c2[2 H]
IUPAC Name 1,2,3,4,5-pentadeuterio-6-[1,1,2,2-tetradeuterio-2-(2,3,4,5,6-pentadeuteriophenyl)ethyl]benzene
Molecular Weight (g/mol) 196.3472
Formula Weight 196.1974 g/mol
Percent Purity 98 atom % D, min 98% Chemical Purity