Learn More
1,2-Dimethoxy-d6-benzene, CDN
Specifications
Specifications
| Chemical Name or Material | Catechol Dimethylether-d6 |
| CAS | 24658-24-0 |
| Molecular Formula | C8 2H6 H4 O2 |
| Quantity | 1 g |
| InChI Formula | InChI=1 S/C8H10O2/c1-9-7-5-3-4-6-8(7)10-2/h3-6 H,1-2H3/i1D3,2D3 |
| SMILES | [2 H]C([2 H])([2 H])Oc1ccccc1OC([2 H])([2 H])[2 H] |
| IUPAC Name | 1,2-bis(trideuteriomethoxy)benzene |
| Molecular Weight (g/mol) | 144.2 |
| Formula Weight | 144.1057 g/mol |
| Percent Purity | 99 atom % D, min 98% Chemical Purity |