Learn More
1,2-Dihydroxynaphthalene, TRC
Product Code. 30386695
Change view
Click to view available options
Quantity:
50 mg
100 mg
500 mg
Unit Size:
100mg
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30386695 500 mg 500mg
30374454 50 mg -
30379937 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30386695 Supplier Toronto Research Chemicals Supplier No. TRCD454168500MG
 Restricted Item
Estimated Shipment: 27-08-2026
Add to basket
Add to basket
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 574-00-5
Molecular Formula C10H8O2
Molecular Weight (g/mol) 160.17
Synonym 1,2-Naphthalenediol, 1,2-Naphthoquinol, 1-Hydroxy-2-naphthol, NSC 401609
SMILES Oc1ccc2ccccc2c1O
Specifications
Quantity 500 mg
InChI Formula InChI=1S/C10H8O2/c11-9-6-5-7-3-1-2-4-8(7)10(9)12/h1-6,11-12H
Formula Weight 160.0524
Chemical Name or Material 1,2-Dihydroxynaphthalene